Order Entry
ContactUsLinkComponent
SPATULA, TAPERED MICRO
Spatulas, Tapered, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
# de catalogue 80071-668
Fournisseur:  Chemglass
Small Business Enterprise
SPATULA, TAPERED MICRO
Spatulas, Tapered, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
# de catalogue 80071-668
Fournisseur:  Chemglass
Small Business Enterprise

Spécifications

  • Length:
    20.3 cm (8")
  • Description:
    Stainless steel spatula, #304, non-magnetic, tapered end
  • Cat. no.:
    80071-668
  • Supplier No.:
    CG-1982-11

Spécifications

A propos de cet article

Spatulas are made of non-magnetic stainless steel and feature a mirror finish.

  • Spatula will pass through a 14/10 or 14/20 outer joint
  • One end is rounded, the other is tapered to ¹/₈" (3.3 mm)

Both ends are approx 8 mm (⁵/₁₆") wide × 51 mm (2") in length.

Customers Who Bought This Also Bought