ContactUsLinkComponent
SPATULA, ROUNDED MICRO
Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Catalog # 80071-664
Supplier:  Chemglass
Small Business Enterprise
SPATULA, ROUNDED MICRO
Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Catalog # 80071-664
Supplier:  Chemglass
Small Business Enterprise
Specifications
  • Description:
    Stainless steel spatula, #304, non-magnetic, rounded ends
  • Length:
    20.3 cm (8")
  • Cat. no.:
    80071-664
  • Supplier No.:
    CG-1981-10
Specifications
About this item
Spatulas are made of non-magnetic stainless steel and feature a mirror finish.
  • Spatula will pass through a 14/10 or 14/20 outer joint
  • Both ends are rounded and are approximately ⁵/₁₆" (8 mm) wide × 2" (51 mm) in length
  • Non-magnetic
Customers Who Bought This Also Bought