Airfree® Schlenk Stoppers, O-Ring, Chemglass
# de catalogue 80064-394
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Airfree® Schlenk Stopper, #15 O-Ring Joint
- Cat. no.:80064-394
- Supplier No.:AF-0210-01
Spécifications
A propos de cet article
For use with the Double Bank, Inert Gas or Single Bank, Inert Gas manifolds or any Airfree® component with an O-Ring as a closure.
Delivery: Supplied complete with one Viton O-Ring.
Customers Who Bought This Also Bought