ContactUsLinkComponent
STOPPER, O-RING, AIRFREE
Airfree® Schlenk Stoppers, O-Ring, Chemglass
# de catalogue 80064-394
Fournisseur:  Chemglass
Small Business Enterprise
STOPPER, O-RING, AIRFREE
Airfree® Schlenk Stoppers, O-Ring, Chemglass
# de catalogue 80064-394
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Description:
    Airfree® Schlenk Stopper, #15 O-Ring Joint
  • Cat. no.:
    80064-394
  • Supplier No.:
    AF-0210-01
Spécifications
A propos de cet article
For use with the Double Bank, Inert Gas or Single Bank, Inert Gas manifolds or any Airfree® component with an O-Ring as a closure.
Delivery: Supplied complete with one Viton O-Ring.
Customers Who Bought This Also Bought