Fritted Filter Discs, Quartz, Chemglass
# de catalogue 80063-702
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Ø:30 mm
- Porosity:Coarse
- Supplier No.:CGQ-0207-14
Spécifications
A propos de cet article
Porous fused quartz discs.
Porosity sizes differ from porosities of borosilicate discs.
Customers Who Bought This Also Bought