ContactUsLinkComponent
QUARTZ FRITTED DISC
Fritted Filter Discs, Quartz, Chemglass
# de catalogue 80063-702
Fournisseur:  Chemglass
Small Business Enterprise
QUARTZ FRITTED DISC
Fritted Filter Discs, Quartz, Chemglass
# de catalogue 80063-702
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Ø:
    30 mm
  • Porosity:
    Coarse
  • Supplier No.:
    CGQ-0207-14
Spécifications
A propos de cet article
Porous fused quartz discs.
Porosity sizes differ from porosities of borosilicate discs.
Customers Who Bought This Also Bought