Spécifications
- Capacity:2 cm³
- Subdivisions:1/10 cc
- Supplier No.:CG-3070-01
Spécifications
A propos de cet article
Borosilicate glass syringes have interchangeable barrels and plungers
Colored plunger ends identify the capacity of the syringe. Flanges are flat on two sides to prevent syringe from rolling. All sizes of syringes are equipped with a metal luer lock and permanently fused graduations