Magnetic Stir Bar Retrivers, Chemglass
# de catalogue 80062-182
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Coating:Polyethylene
- Ø:9.5 mm (3/8 ")
- Length:457 mm (18")
- Description:Stir Bar Retriever
- Cat. no.:80062-182
- Supplier No.:CG-2015-02
Spécifications
A propos de cet article
The 3/8" diameter pick up rod features a powerful Alnico magnet in one end, and a hang-up grip on the other
Available in either 12" or 18" lengths.
Customers Who Bought This Also Bought