ContactUsLinkComponent
MELTING POINT TUBE, CLOSED ONE END
Melting Point Tubes, Chemglass
# de catalogue 80061-552
Fournisseur:  Chemglass
Small Business Enterprise
MELTING POINT TUBE, CLOSED ONE END
Melting Point Tubes, Chemglass
# de catalogue 80061-552
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Length:
    90 mm
  • Description:
    Melting Point Tubes
  • Cat. no.:
    80061-552
  • Supplier No.:
    CG-1841-01
Spécifications
A propos de cet article
Tubes are manufactured from borosilicate glass and packaged 100 tubes per vial. Closed at one end.
Tube 80061-554 has a smaller I.D. to conform to U.S.P XXI requirements.
Customers Who Bought This Also Bought