Melting Point Tubes, Chemglass
# de catalogue 80061-552
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Length:90 mm
- Description:Melting Point Tubes
- Cat. no.:80061-552
- Supplier No.:CG-1841-01
Spécifications
A propos de cet article
Tubes are manufactured from borosilicate glass and packaged 100 tubes per vial. Closed at one end.
Tube 80061-554 has a smaller I.D. to conform to U.S.P XXI requirements.
Customers Who Bought This Also Bought