Accessories for Reaction Vessels, Chemglass
# de catalogue 80061-152
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Flange, for Reaction Kettle, with O-Ring Groove
- Cat. no.:80061-152
- Flange I.D.:120 mm
- Supplier No.:CG-147-03
Spécifications
A propos de cet article
Flange is formed with a groove in the flat portion of the mating surface to accept an O-Ring.
It also has a ground surface, permitting assembly with either the CG-148 flat flange or CG-149 lid blank using grease.
In the greased application, the O-Ring groove prevents grease from entering the reaction kettle and causing possible contamination.
Outer taper of flange is formed at a 45° angle to accept the CG-141 clamp.
Customers Who Bought This Also Bought