Safety Wash Bottles, Chemglass
# de catalogue 80060-416
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Capacity:500 ml (16.9 oz.)
- Description:Wash Bottle, Ethanol
- Material:Polyethylene
- Cat. no.:80060-416
- Supplier No.:CG-1130-06
Spécifications
A propos de cet article
Easy-to-read, low-density polyethylene dispensing bottles are break-resistant and include DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ information on their exterior.
Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.
Wash bottles are intended for short-term dispensing use.
Customers Who Bought This Also Bought