ContactUsLinkComponent
Hydrion Insta-Check 0-13 pH Paper Dispenser
Hydrion® Double Roll pH Paper, Micro Essential Laboratory
# de catalogue 73450-010
Fournisseur:  Micro Essential
Hydrion Insta-Check 0-13 pH Paper Dispenser
Hydrion® Double Roll pH Paper, Micro Essential Laboratory
# de catalogue 73450-010
Fournisseur:  Micro Essential
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    pH Paper Dispenser, 0.0 - 13.0 pH Double Roll
  • Supplier No.:
    193
Spécifications
A propos de cet article
Hydrion® Double Roll pH Paper set the standard for quality, accuracy, and reliability.
Double Roll pH Papers consist of two rolls of pH papers packaged in a protective plastic dispenser with stainless steel cutoff lips and two matching color charts
Double Roll pH Wide Range Papers have a clear color match at each full pH unit: [0.0][1.0][2.0][3.0][4.0][5.0][6.0][7.0][8.0][9.0][10.0][11.0][12.0][13.0][14.0]
pH color changes are distinct and easily matched to the color chart.
Ordering information: Each carton (case) contains 10 dispensers, each consisting of two (2) 15-foot rolls of test paper and matching color charts. Provides for approximately 2000 tests.
Customers Who Bought This Also Bought