HydroLogix HLT 5000 Pipette Tips, Molecular BioProducts
# de catalogue 72830-734
Fournisseur: Thermo Scientific
Spécifications
- Volume:5000 µl
- Tip Color:Clear
- Packed:Bulk, 250 Tips/bag
- Tip Style:Standard Tips
- Supplier No.:3897-11
Spécifications
A propos de cet article
These non-filtered tips are made of polypropylene.
- Clear
- MBP clear HydroLogix tip
- Non-sterile
Customers Who Bought This Also Bought