ContactUsLinkComponent
TIP HLT BLUE 1000UL PK1000
HydroLogix HLT Blue Pipette Tips, Molecular BioProducts
Catalog # 72830-688
Supplier:  Thermo Scientific
   Low Manufacturing Impact
HydroLogix HLT Blue Pipette Tips, Molecular BioProducts
Catalog # 72830-688
Supplier:  Thermo Scientific
   Low Manufacturing Impact
Specifications
  • Volume:
    1000 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Tip Color:
    Blue
  • Packed:
    Bulk, non-sterile, 1000 Tips/bag
  • Cat. no.:
    72830-688
  • Tip Style:
    Standard Tips
  • Supplier No.:
    3950-11
Specifications
About this item
These non-filtered pipette tips utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipette tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
Customers Who Bought This Also Bought