Pure™ HLT Yellow Pipette Tips, Molecular BioProducts
# de catalogue 72830-662
Fournisseur: Thermo Scientific
Spécifications
- Volume:200 µl
- Tip Color:Yellow
- Packed:Bulk, 1000 Tips/bag
- Tip Style:Standard Tips, Thin Wall
- Supplier No.:3960
Spécifications
A propos de cet article
All HydroLogix tubes feature secure sealing caps to minimize sample evaporation during thermal cycling.
- Reduce strains and injury
- Secure and consistent seals
- Less ejection force
- Available in low retention polymer
All HydroLogix tubes are compatible with major brand thermocyclers.
Customers Who Bought This Also Bought