Automation Pipette Tips for Caliper Workstations, Thermo Scientific
# de catalogue 72830-602
Fournisseur: Thermo Scientific
Spécifications
- Volume:50 µl
- Tip Color:Clear
- Packed:96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
- Tip Style:Standard
- For Use With:Caliper automated workstations
- Supplier No.:920-262
Spécifications
A propos de cet article
Thermo scientific automation pipette tips made to fit the Caliper RapidPlate, Presto, Sciclone, Zephry, and Allegro Automated Workstations.
It provides a solution for your Beckman robotic pipette tips. These automation pipette tips have been designed, validated, and tested on a Caliper robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Caliper workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Customers Who Bought This Also Bought