Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
Catalogue # 72830-422
Fournisseur: Thermo Scientific
Spécifications
- Volume:0.1 - 10 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. no.:72830-422
- Sterile:+
- Tip Style:Pure 10
- Supplier No.:3501-05
Spécifications
À propos de cet article
Molecular BioProducts low retention pipette tips are specifically designed for eliminating sample retention within the pipette tip.
All Pure tips are racked, pre-sterilized, and certified free of RNase, DNase, DNA, and pyrogen.
The inner surface of the tip is extremely hydrophobic, reducing loss of sample from adhesion to the tip interior. Tips are crystal clear to allow easy sample viewing.
Frequently Bought Together