Thermo Scientific™ Automation Tips for Revvity™ Sciclone™ and Zymark™ Liquid Handling Workstations
Catalogue # 72830-242
Fournisseur: Thermo Scientific
Spécifications
- Volume:200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Tip Color:Clear
- Packed:50 racks/case
- Cat. no.:72830-242
- Sterile:-
- For Use With:Revvity™ Sciclone™ and Zymark™ workstations
- Supplier No.:923-262
Spécifications
À propos de cet article
Enhance pipetting performance with Thermo Scientific™ automation tips for Revvity™ Sciclone™ and Zymark™ liquid handling workstations. These tips undergo a rigorous 15-point quality control program, ensuring consistency, superior tip straightness, and a low coefficient of variation for accurate and precise performance.
Optionally available as sterile and filtered
Optional sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Lot-tested during production on actual workstation for which they are created
Low percentage coefficient of variation (%CV)
Racked tips
Thermo Scientific automation tips ensure precision and reliability for Sciclone™ and Zephyr™ workstations, optimizing accuracy and efficiency.
Frequently Bought Together