ContactUsLinkComponent
Thermo Scientific™ Automation Tips for Revvity™ Sciclone™ and Zymark™ Liquid Handling Workstations
Catalogue # 72830-242
Fournisseur:  Thermo Scientific
   Low Manufacturing Impact
Thermo Scientific™ Automation Tips for Revvity™ Sciclone™ and Zymark™ Liquid Handling Workstations
Catalogue # 72830-242
Fournisseur:  Thermo Scientific
Supplier Number:  923-262
   Low Manufacturing Impact
Spécifications
  • Volume:
    200 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Tip Color:
    Clear
  • Packed:
    50 racks/case
  • Cat. no.:
    72830-242
  • Sterile:
    -
  • For Use With:
    Revvity™ Sciclone™ and Zymark™ workstations
  • Supplier No.:
    923-262
Spécifications
À propos de cet article
Enhance pipetting performance with Thermo Scientific™ automation tips for Revvity™ Sciclone™ and Zymark™ liquid handling workstations. These tips undergo a rigorous 15-point quality control program, ensuring consistency, superior tip straightness, and a low coefficient of variation for accurate and precise performance.
  • Optionally available as sterile and filtered
    Optional sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
    Lot-tested during production on actual workstation for which they are created
    Low percentage coefficient of variation (%CV)
    Racked tips
Thermo Scientific automation tips ensure precision and reliability for Sciclone™ and Zephyr™ workstations, optimizing accuracy and efficiency.
Frequently Bought Together