Clear Pipette Tips for Zymark® Workstations, Molecular BioProducts
# de catalogue 72830-240
Fournisseur: Thermo Scientific
Spécifications
- Volume:200 µl
- Sterile:+
- Supplier No.:923-261
Spécifications
A propos de cet article
Optimized for Zymark®-style workstations.
- ART tips are pre-sterilized and certified RNase-, DNase-, DNA-, and pyrogen-free
MBP® BioRobotix® brand tips are tested for compatibility on the actual machines for which they are designed and are available with ART® barriers for guaranteed protection against cross contamination. ART® self-sealing barriers trap contaminants, such as aerosols and liquids, keeping them out of pipettors and samples. This allows the user to conduct PCR procedures and pipette radioactive or infectious samples.
Tips can be used with the Zymark RapidPlate® 96, Presto®, and SciClone™ (200 µl only) workstations. Tips are long enough for use with deep well plates and tubes.
Ordering information: Tips are packaged 96 per tray, 10 trays per pack.
Customers Who Bought This Also Bought