ContactUsLinkComponent
Hydrion® Single Roll Dispenser, pH 1.0 to 12.0, Micro Essential Laboratory®
Catalogue # 71002-042
Fournisseur:  Micro Essential
undefined
Hydrion® Single Roll Dispenser, pH 1.0 to 12.0, Micro Essential Laboratory®
Catalogue # 71002-042
Fournisseur:  Micro Essential
Supplier Number:  94
Spécifications
  • Description:
    Wide Range pH Test Paper
  • Cat. no.:
    71002-042
  • pH Range:
    1.0 - 14.0
  • Supplier No.:
    94
Spécifications
À propos de cet article
Hydrion® Insta-Chek pH paper is an excellent direct reading pH paper with the widest range available in a single paper.
  • Simple and single color match
  • Distinct and easily matched color separations
  • Accurate, reproducible readings with an immediate response
General purpose pH papers has a distinct color match at full pH units.
Packaging: Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart. Provides for approximately 1000 tests.
Frequently Bought Together