Order Entry
ContactUsLinkComponent
Flask
Flask, Shaker, Deep Baffles, Ace Glass Incorporated
Catalog # 71000-426
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Flask
Flask, Shaker, Deep Baffles, Ace Glass Incorporated
Catalog # 71000-426
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Cap size:
    #6
  • Capacity:
    250 ml
  • Description:
    Shaker Flask, Deep Baffles with Beaded Neck
  • Cat. no.:
    71000-426
  • O.D.×H:
    85 x 130 mm
  • Supplier No.:
    3890-04

Specifications

About this item

This shaker flask has three extra deep baffles and a beaded neck

It is used for converting homogenous tissue samples into cell suspensions by digestion of connective tissues. Fabricated from borosilicate glass.

Ordering information: Rubber stopper not included.
Customers Who Bought This Also Bought