Order Entry
ContactUsLinkComponent
2Mm 2-Way PTFE Stopcock Plug
Connectors, with PTFE Plug Valve, Ace Glass Incorporated
Catalog # 71000-282
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Connectors, with PTFE Plug Valve, Ace Glass Incorporated
Catalog # 71000-282
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Connectors with Two-Way Valve
  • Cat. no.:
    71000-282
  • Bore Size:
    2 mm
  • Supplier No.:
    12611-04

Specifications

About this item

Polypropylene connectors, two-way or three-way, with PTFE valve and serrated tubulations

Takes 6.4 mm to 9.5 mm I.D. tubing.

Frequently Bought Together