Safety Wash Bottle, Ace Glass
# de catalogue 71000-206
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Toluene, Safety Labeled Wash Bottles
- Cat. no.:71000-206
- Supplier No.:12464-16
Spécifications
A propos de cet article
These wide mouth safety wash bottles are guaranteed to be leak-proof.
They are made of flexible, impact-resistant polyethylene with color-coded polyallomer closure and spout. Standard 'diamond alert' symbols indicate what type hazard the contents may present. Available in 500 ml (16 oz.) size only.
Customers Who Bought This Also Bought