SP Bel-Art Poxygrid Bag Holders, Bel-Art Products, a part of SP
Catalog # 70680-356
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:37 cm (14.6")
- Description:Bag holder medium for bag size 24x30"
- Height:60 cm (23.5")
- Cat. no.:70680-356
- Supplier No.:131880000
Specifications
About this item
Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place.
- Bright biohazard orange colored epoxy-coated steel wire
- Chemical- and corrosion-resistant
It will withstand a temperature range of –223 °C (–369 °F) to 121 °C (250 °F) and is autoclavable.
Frequently Bought Together