SP Bel-Art Biohazard Disposal Bags, Plain or Printed, Bel-Art Products, a part of SP
Catalog # 70680-346
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:1 - 3 gal.
- Color:Red with Biohazard Logo
- Thickness:1.5 mil.
- W×L:305×610 mm (12×24")
- Supplier No.:131600009
Specifications
About this item
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
- Economical
- Available in a variety of labeling options--plain or printed
- High temperature autoclavable at a maximum of 135 °C (275 °F)
Available plain, non-printed or with biohazard label and indicator patch. Plain, non-printed bags can also be used as conventional trash can liners.
Certifications: Meet or exceed the ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests.
Frequently Bought Together