ContactUsLinkComponent
Delrin* Screw Clamp
SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
# de catalogue 70680-322
Delrin* Screw Clamp
SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
# de catalogue 70680-322
Spécifications
  • Cat. no.:
    70680-322
  • Fits Tubing O.D.:
    max. 14 mm (¹/₂")
  • Supplier No.:
    182120000
Spécifications
A propos de cet article
Offers on/off and fine control so accurate that it replaces stopcocks in many applications.
  • Molded acetal plastic construction
  • Very fine, accurate control
  • Autoclavable
  • Resistant to acids, alkalis, and organic solvents
Can be installed without disconnecting tubing or interrupting operation. Molded of Acetal plastic, the clamp is autoclavable and resistant to acids, alkalis and organic solvents. Accepts flexible tubing up to 14 mm (½") O.D.
Customers Who Bought This Also Bought