SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
# de catalogue 70680-270
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Size:76 cm (30") wide x 15.2 meter (50 ft)
- Cat. no.:70680-270
- Supplier No.:246790000
Spécifications
A propos de cet article
Clear polyethylene ribbed matting can be used for lining laboratory tables, shelves, trays, etc.
- Clear polyethylene construction
- Ribbed design
- Easily trimmed to fit laboratory surfaces
It is easily trimmed with scissors and also works well under drums or carboys. 0.25 mm (0.010") with 0.7 mm (0.030") thick ribs.
Customers Who Bought This Also Bought