ContactUsLinkComponent
ADAPTER NOVA SQUEEGEE SS 12IN
NovaSqueegee™ Cleanroom, Micronova
# de catalogue 68400-536
Fournisseur:  Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
ADAPTER NOVA SQUEEGEE SS 12IN
NovaSqueegee™ Cleanroom, Micronova
# de catalogue 68400-536
Fournisseur:  Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Description:
    Stainless steel adapter
  • Width:
    30.5 cm (12")
  • Supplier No.:
    QDNS-12
Spécifications
A propos de cet article
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors, and walls in the cleanroom. Optional PVA blades are fast-drying, low-linting, and low in particulates with a 600% absorbency rate; ideal for electronic and semiconductor cleanrooms.
  • Designed to clean windows, screens, floors, and walls in the cleanroom
  • Made with 304 electropolished stainless steel and Derlin® resin
  • Rubber replacement blades available
  • PVA replacement blades available for absorbency
  • Quick disconnect handle attachment
  • Entire unit is autoclavable
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a Quick Disconnect handle attachment for optional use with the SSU series handles for extended reach.
Customers Who Bought This Also Bought