Order Entry
ContactUsLinkComponent
NovaPoly* Slip Cover
T-Mop™ Cleanroom Mop Covers, Micronova
# de catalogue 68400-404
Fournisseur:  Micronova
   Waste Reducing
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
NovaPoly* Slip Cover
T-Mop™ Cleanroom Mop Covers, Micronova
# de catalogue 68400-404
Fournisseur:  Micronova
   Waste Reducing
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Spécifications

  • Description:
    NovaPoly™ mop cover
  • Environmentally Preferable:
    Waste Reducing
  • Material:
    Polyester, smooth knit
  • Length:
    45.7 cm (18")
  • Supplier No.:
    T6-18

Spécifications

A propos de cet article

A series of rounded T-mop covers with snaps that easily roll down walls, as well as clean awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialized cleaning.

  • Low profile, rounded covered sponges
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces, and under equipment
  • Autoclavable
  • Holds up to strong disinfectants

Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester for removing residues and dispersing disinfectant
MegaTex™ - 100% non-woven polyamide is absorbent with a mildly abrasive surface
Microfiber - 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
PolyMesh™ - 100% polyester with a layer of mesh provides a mild scrubbing surface
NovaLite™ - absorbent synthetic chamois provides a fine clean and finish

The Micronova T-Mop™ Cleanroom Mop Covers fit onto the 180° swivel TMop™ adapter. Available with an optional thicker foam layer to hold more disinfectant and facilitate wringing. TMC6-14 is an elastic single-ply slipcover, a low cost option for smaller areas.

Ordering information: All components must be ordered separately. For additional universal handle lengths, see universal mop handles from Micronova.
Customers Who Bought This Also Bought