ContactUsLinkComponent
NovaDyne* General Purpose Soap
Hand care products, Micronova
# de catalogue 68400-374
Fournisseur:  Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
NovaDyne* General Purpose Soap
Hand care products, Micronova
# de catalogue 68400-374
Fournisseur:  Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Capacity:
    500 ml (16.9 oz.)
  • Description:
    Novadyne™ general purpose hand soap
  • Supplier No.:
    IC-110
Spécifications
A propos de cet article
The Micronova Hand Care Products include a range of soaps and sanitizers that are formulated with highly effective, non-irritating ingredients that are ideal for frequent use in clean and controlled environments.
  • Saddle pump bottles can be used as standalone dispensers
  • Saddle pump bottles can also be used with VWR® MicroDispenser™ Touch-Free Dispenser to ensure uncontaminated results
NovaClenz™ Instant Hand Sanitizer is made with 62% Ethanol to help prevent viral transfer. Effective against bacteria, including pseudomonas, salmonella, and staphylococcus. Effective in reducing nosocomial infections. Fortified with emollients to moisturize hands.
Hy-G-Clenz™ Antimicrobial Hand Soap is made with 0.13% Benzalkonium Chloride and natural emollients. Effective against a wide range of both gram-positive and gram-negative bacteria. Persistent activity up to three hours. The soap is pH balanced and contains natural moisturizers, ideal for frequent hand washing. Meets OSHA standards.
AquaHol™ Sterile Isopropyl Alcohol Solution is made with 70% sterile IPA in 30% Wifi quality water, and meets USP requirements. Terminally filtered to 0.1 microns. Gamma Irradiated. Ideal for use over cleanroom gloves.
NovaDyne™ General-Purpose Hand Soap is a pH balanced, gentle lotion hand soap. Made with built-in moisturizers/skin emollients, it is ideal for frequent hand washing.
Certifications: Compliant with OSHA standards.
Ordering information: Pump bottles can be used as stand-alone dispensers or with the VWR® MicroDispenser touch-free soap dispenser.
Customers Who Bought This Also Bought