ContactUsLinkComponent
REFRACTOMETER SALT 0.28% NACL
Salt Refractometer, 0 to 28%, Sper Scientific
Catalog # 66214-986
Supplier:  Sper Scientific
REFRACTOMETER SALT 0.28% NACL
Salt Refractometer, 0 to 28%, Sper Scientific
Catalog # 66214-986
Supplier:  Sper Scientific
Specifications
  • Description:
    Salt Refractometer 0–28%
  • Cat. no.:
    66214-986
  • Supplier No.:
    300006
Specifications
About this item
Refractometer measures salt content in sea water, pickling-brines, and other applications
While the scale is calibrated for sodium chloride, it can also be used for calcium chloride, barium cobalt, magnesium, cesium, potassium, sodium chloride, cupric sulfate, potassium iodide, citric acid, acetic acid, and vinegar. To operate the device, simply place a drop or two of sample on the prism and read the results.
Ordering information: Refractometer is supplied with a hard carrying case, calibration tool, and 5-year warranty.
Customers Who Bought This Also Bought