ContactUsLinkComponent
Vaccu-Pette/96* Pipetting Device
SP Bel-Art Vaccu-Pette 96-well Format Pipetting Device, Bel-Art Products, a part of SP
# de catalogue 66190-080
Vaccu-Pette/96* Pipetting Device
SP Bel-Art Vaccu-Pette 96-well Format Pipetting Device, Bel-Art Products, a part of SP
# de catalogue 66190-080
Spécifications
  • Description:
    Pipetting Device, Autoclavable, Reusable Polycarbonate
  • Cat. no.:
    66190-080
  • Dimensions:
    12.7L x 8.6W x 3.7H cm
  • Supplier No.:
    378760001
Spécifications
A propos de cet article
This pipetting device can aspirate or dispense 96 wells in less than 10 seconds.
  • Transparent polycarbonate construction
  • Available in sterile/disposable or autoclavable/reusable models
  • Ideal for efficient washing, dispensing, or aspirating of wells
It speeds up repetitive washing, dispensing, or aspirating of wells by working simultaneously. It has a vacuum outlet at one end, and a short length of 3.2 mm I.D. (1/8") tubing is supplied for syringe connection.
Sterile/disposable model is molded of clear, disposable plastic that can be used only once and cannot be autoclaved or gas sterilized. Autoclavable/Reusable model is molded of polycarbonate, and must be sterilized by autoclaving at no more than 121 °C for 20 minutes at 15 psi prior to use in order to avoid contamination. It cannot be gas sterilized due to the possibility of residual trapped gases.
Simply connect a disposable plastic syringe and withdraw the plunger to transfer liquid. Excellent for simultaneously changing media or for the addition/removal of reagents from a 96-well plate.
Ordering information: Requires a 30 ml or larger disposable sterile syringe (not included).
Delivery: Complete with a 7.6 cm (3") long piece of 3.2 mm (1/8") tubing for syringe connection. The autoclavable, reusable polycarbonate pipetting device is supplied in a sealed pack with tubing enclosed.
Customers Who Bought This Also Bought