Spécifications
- Description:Pipetting Device, Autoclavable, Reusable Polycarbonate
- Cat. no.:66190-080
- Dimensions:12.7L x 8.6W x 3.7H cm
- Supplier No.:378760001
Spécifications
A propos de cet article
This pipetting device can aspirate or dispense 96 wells in less than 10 seconds.
- Transparent polycarbonate construction
- Available in sterile/disposable or autoclavable/reusable models
- Ideal for efficient washing, dispensing, or aspirating of wells
It speeds up repetitive washing, dispensing, or aspirating of wells by working simultaneously. It has a vacuum outlet at one end, and a short length of 3.2 mm I.D. (1/8") tubing is supplied for syringe connection.
Sterile/disposable model is molded of clear, disposable plastic that can be used only once and cannot be autoclaved or gas sterilized. Autoclavable/Reusable model is molded of polycarbonate, and must be sterilized by autoclaving at no more than 121 °C for 20 minutes at 15 psi prior to use in order to avoid contamination. It cannot be gas sterilized due to the possibility of residual trapped gases.
Simply connect a disposable plastic syringe and withdraw the plunger to transfer liquid. Excellent for simultaneously changing media or for the addition/removal of reagents from a 96-well plate.