Specifications
- Volume:3 ml
- Cat. no.:66064-770
- Supplier No.:S7515-3
Specifications
About this item
All-plastic syringes are free of rubber components or lubricants
These syringes are preferred for use with Target™ brand syringe filters. Polyethylene barrels and polypropylene plungers offer wide solvent compatibility, low background extractables and the convenience of a disposable product. No silicone or oil lubricant is required in the barrel.