ContactUsLinkComponent
Luer Lok Plastic Disposable Syringes, Capacity: 3 ml
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Catalog # 66064-770
Supplier:  Thermo Scientific
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Catalog # 66064-770
Supplier:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    3 ml
  • Cat. no.:
    66064-770
  • Supplier No.:
    S7515-3
Specifications
About this item
All-plastic syringes are free of rubber components or lubricants
These syringes are preferred for use with Target™ brand syringe filters. Polyethylene barrels and polypropylene plungers offer wide solvent compatibility, low background extractables and the convenience of a disposable product. No silicone or oil lubricant is required in the barrel.
Frequently Bought Together