Spécifications
- Volume:5 ml
- Supplier No.:S7510-5
Spécifications
A propos de cet article
Luer-Slip syringes feature polyethylene barrels and polypropylene plungers
For use in all syringe filter applications. Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets.