Specifications
- Volume:20 ml
- Cat. no.:66064-756
- Supplier No.:S7510-20
Specifications
About this item
Luer-Slip syringes feature polyethylene barrels and polypropylene plungers
For use in all syringe filter applications. Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets.