Caps for 13-425 Screw-Thread Vials
# de catalogue 66030-392
Fournisseur: Thermo Scientific
Spécifications
- Cap Size:13-425
- Cap Type:Polypropylene
- Septum:None
- Description:Open-top screw cap
- Cap Color:Black
- Supplier No.:C4015-1A
Spécifications
A propos de cet article
Screw thread caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.
- Polypropylene caps are chemically inert and suitable for most chromatography applications
- Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
- Caps are bonded septa resist dislodging during injection when using large diameter blunt needles
Convenient pre-assembled caps and septa minimize contamination from handling.
Customers Who Bought This Also Bought