ContactUsLinkComponent
Black Polypropylene Screw Cap
Caps for 13-425 Screw-Thread Vials
# de catalogue 66030-392
Fournisseur:  Thermo Scientific
Caps for 13-425 Screw-Thread Vials
# de catalogue 66030-392
Fournisseur:  Thermo Scientific
Spécifications
  • Cap Size:
    13-425
  • Cap Type:
    Polypropylene
  • Septum:
    None
  • Description:
    Open-top screw cap
  • Cap Color:
    Black
  • Supplier No.:
    C4015-1A
Spécifications
A propos de cet article
Screw thread caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
  • Caps are bonded septa resist dislodging during injection when using large diameter blunt needles
Convenient pre-assembled caps and septa minimize contamination from handling.
Customers Who Bought This Also Bought