Septa for 13-425 Screw-Thread Vials, Thermo Scientific
# de catalogue 66030-390
Fournisseur: Thermo Scientific
Spécifications
- Septum:Red PTFE/white silicone
- Description:Septa
- Cat. no.:66030-390
- Supplier No.:C4015-60
Spécifications
A propos de cet article
Autosampler vial septa, 13 mm for use with 13-425 threaded caps or 13 mm snap/crimp caps.
- PTFE disk septa are chemically inert
- PFTE/red rubber septa are economical for many GC applications
- PTFE/silicone septa are most popular for HPLC and IC applications
- Pre-slit septa are easier to pierce with thin gauge needles
Septa are available in PTFE/ red rubber, PTFE/silicone or red PTFE/white silicone/red PTFE to suit a variety of applications.
Customers Who Bought This Also Bought