ContactUsLinkComponent
Septa for 13mm Screw Thread Caps, Red PTFE / White Silicone, 0.060" Septa
Septa for 13-425 Screw-Thread Vials, Thermo Scientific
# de catalogue 66030-390
Fournisseur:  Thermo Scientific
Septa for 13-425 Screw-Thread Vials, Thermo Scientific
# de catalogue 66030-390
Fournisseur:  Thermo Scientific
Spécifications
  • Septum:
    Red PTFE/white silicone
  • Description:
    Septa
  • Cat. no.:
    66030-390
  • Supplier No.:
    C4015-60
Spécifications
A propos de cet article
Autosampler vial septa, 13 mm for use with 13-425 threaded caps or 13 mm snap/crimp caps.
  • PTFE disk septa are chemically inert
  • PFTE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles
Septa are available in PTFE/ red rubber, PTFE/silicone or red PTFE/white silicone/red PTFE to suit a variety of applications.
Customers Who Bought This Also Bought