Order Entry
ContactUsLinkComponent
VWR* Vial
VWR® Vials, Borosilicate Glass, with Phenolic Screw Cap
# de catalogue 66012-066
Fournisseur:  VWR International
VWR* Vial
VWR® Vials, Borosilicate Glass, with Phenolic Screw Cap
# de catalogue 66012-066
Fournisseur:  VWR International

Spécifications

  • Cap included:
    Yes
  • Cap Size:
    24-400
  • Capacity:
    35.12 ml
  • Cat. no.:
    66012-066
  • Diameter ×H:
    28×95 mm

Spécifications

A propos de cet article

These general-purpose vials are available in either a short form or capsule style.

  • Borosilicate glass fabrication
  • Short form or capsule style
  • Cap is flush to vial shoulder
  • Black phenolic molded screw cap
  • Polyvinyl-faced pulp liner
  • General-purpose use

Vials have black phenolic molded screw cap with polyvinyl-faced pulp liner. The cap on the vial is the same diameter as the body, fitting flush to the shoulder.

Customers Who Bought This Also Bought