Gray Butyl Rubber Stoppers, Kimble Chase
# de catalogue 66010-865
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Gray Butyl Rubber Stopper for Lyophilization, 2 Legs
- Cat. no.:66010-865
- Fits Neck Size:13 mm
- Supplier No.:73828-13
Spécifications
A propos de cet article
Lyophilization style, slotted stopper for aluminum seals.
- Two-leg style reduces possibility of legs sticking together
Gray high grade butyl rubber.
Customers Who Bought This Also Bought