ContactUsLinkComponent
Gray Butyl Rubber Stopper
Gray Butyl Rubber Stoppers, Kimble Chase
# de catalogue 66010-865
Gray Butyl Rubber Stopper
Gray Butyl Rubber Stoppers, Kimble Chase
# de catalogue 66010-865
Spécifications
  • Description:
    Gray Butyl Rubber Stopper for Lyophilization, 2 Legs
  • Cat. no.:
    66010-865
  • Fits Neck Size:
    13 mm
  • Supplier No.:
    73828-13
Spécifications
A propos de cet article
Lyophilization style, slotted stopper for aluminum seals.
  • Two-leg style reduces possibility of legs sticking together
Gray high grade butyl rubber.
Customers Who Bought This Also Bought