SP Bel-Art Acetal Plastic Tubing Clamps, Autoclavable, Bel-Art Products, a part of SP
Catalogue # 63022-407
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:12-Position Maxi Clamp
- Cat. no.:63022-407
- Supplier No.:182290000
Spécifications
À propos de cet article
These acetal clamps will not distort, rust, or corrode.
- Acetal plastic construction
- Multiple models available with varying position options
- Multi-position clamps provide full range of control over closure
Single Position Mini Clamp: Single-position, on-off clamp for use with thin-walled tubing under 4.7 mm O.D
12-Position Maxi Clamp: 12-position ratchet control from full open to complete closure. For tubing up to 19 mm O.D. (3/4").
15-Position Mid-Range Clamp: 15-position stepped control throttles flow from full open to complete closure. Fits tubing from 3.2–11 mm O.D. (1/8–7/16").
(3/16"). Gives positive shut-off with finger tip control.
Frequently Bought Together