ContactUsLinkComponent
VWR* Black Vacuum Rubber Tubing
VWR® Vacuum Rubber Tubing, Black
# de catalogue 62994-219
Fournisseur:  VWR International
Small Business Enterprise
VWR* Black Vacuum Rubber Tubing
VWR® Vacuum Rubber Tubing, Black
# de catalogue 62994-219
Fournisseur:  VWR International
Small Business Enterprise
Spécifications
  • Wall thickness:
    1/4"
  • Cat. no.:
    62994-219
  • Coil Length:
    9¾ ft.
  • I.D.×O.D.:
    ⁵⁄₁₆×¹³⁄₁₆"
Spécifications
A propos de cet article
Tubing has especially heavy outer wall, which makes it suitable for vacuum use.
  • Seamless
  • Acid-proof
  • Good flexing qualities at low temperatures
Manufactured from good-quality natural rubber. Tubing also offers high resilience, good tensile strength, and tear resistance. Durometer hardness: Shore A, 50. Tensile strength: 2500 to 3500 psi.
Customers Who Bought This Also Bought