SP Bel-Art Piezoelectric Gas Lighter, Bel-Art Products, a part of SP
# de catalogue 62866-127
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Gas lighter
- Cat. no.:62866-127
- Length:26.5 cm (10.5")
- Weight:88 g (3.1 oz.)
- Supplier No.:150560000
Spécifications
A propos de cet article
Safely light Bunsen burners, gas stoves, propane or oxy-acetylene torches.
- Comfortable plastic grip
- Bush-button trigger creates spark
- No need for batteries or fuel refills
Requiring no batteries or fuel refills, the Piezo Electric Gas Lighter has a comfortable plastic pistol-grip with a push-button trigger that creates an electrical spark (not a flame) at 20 KV with each squeeze.
Customers Who Bought This Also Bought