Nalgene® Tubing Connectors, Polypropylene, Thermo Scientific
Catalog # 62850-060
Supplier: Thermo Fisher Scientific
Specifications
- Tubing I.D.:6.4 mm (¼")
- Cat. no.:62850-060
- Supplier No.:6152-0250
Specifications
About this item
Durable polypropylene connectors are resistant to gases, solvents, and acids
Available in T- and Y-type connectors. Autoclavable.
Customers Who Bought This Also Bought