ContactUsLinkComponent
Nalgene Tubing Connectors, Polypropylene, Thermo Scientific
Nalgene® Tubing Connectors, Polypropylene, Thermo Scientific
Catalog # 62850-060
Nalgene Tubing Connectors, Polypropylene, Thermo Scientific
Nalgene® Tubing Connectors, Polypropylene, Thermo Scientific
Catalog # 62850-060
Specifications
  • Tubing I.D.:
    6.4 mm (¼")
  • Cat. no.:
    62850-060
  • Supplier No.:
    6152-0250
Specifications
About this item
Durable polypropylene connectors are resistant to gases, solvents, and acids
Available in T- and Y-type connectors. Autoclavable.
Customers Who Bought This Also Bought