ContactUsLinkComponent
Nalgene Pans, High-Density Polyethylene, Thermo Scientific
Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
# de catalogue 62662-886
Nalgene Pans, High-Density Polyethylene, Thermo Scientific
Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
# de catalogue 62662-886
Spécifications
  • Dimensions:
    54.3L x 43.5W x 13H cm (213/8 x 171/8 x 51/8")
  • Cat. no.:
    62662-886
  • Supplier No.:
    7120-0020
Spécifications
A propos de cet article
Pans are constructed of high-density polyethylene for rigidity and durability
Use for soaking labware or as an ice bath. With handles for easy carrying. Tapered for nesting.
Customers Who Bought This Also Bought