Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
# de catalogue 62662-875
Fournisseur: Thermo Fisher Scientific
Spécifications
- Dimensions:35.8L x 30.8W x 14.3H cm (141/8 x 121/8 x 55/8")
- Supplier No.:7120-0010
Spécifications
A propos de cet article
Pans are constructed of high-density polyethylene for rigidity and durability
Use for soaking labware or as an ice bath. With handles for easy carrying. Tapered for nesting.
Customers Who Bought This Also Bought