SP Bel-Art Sterilizing Trays and Covers, Polypropylene, Bel-Art Products, a part of SP
# de catalogue 62662-401
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Capacity:5 L (1.3 gal.)
- Cat. no.:62662-401
- L×W×H:30 x 20 x 13 cm (12 x 73/4 x 51/8")
- Supplier No.:162600000
Spécifications
A propos de cet article
Molded polypropylene plastic trays have rounded corners and smooth, tapered sides for stacking.
- Polypropylene construction
- Rounded corners and smooth, tapered sides
- Stackable
- Autoclavable
Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks.
Optional polypropylene covers are 3.2 mm (⅛") thick with a 25.4 mm (1") skirt that fits snugly over the rim of the tray. Trays and lids are steam autoclavable at 121ºC (250ºF).
Ordering information: Trays and covers sold separately.
Customers Who Bought This Also Bought