ContactUsLinkComponent
Nalgene Pans, Polypropylene, Thermo Scientific
Nalgene® Pans, Polypropylene, Thermo Scientific
Catalog # 62662-241
Nalgene Pans, Polypropylene, Thermo Scientific
Nalgene® Pans, Polypropylene, Thermo Scientific
Catalog # 62662-241
Specifications
  • Cat. no.:
    62662-241
  • L×W×H:
    32.4 x 25.7 x 11 cm (123/4 x 10 x 4")
  • Supplier No.:
    6902-5000
Specifications
About this item
Pans are lightweight, durable, chemically inert, and autoclavable
For repeated autoclaving of small instruments and labware. Can also be used for soaking, cleaning, storing, or carrying general lab products. Heavy-duty construction with extra strong rims. Tapered for nesting.
Customers Who Bought This Also Bought