Nalgene® Pans, Polypropylene, Thermo Scientific
# de catalogue 62662-183
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cat. no.:62662-183
- L×W×H:26 x 16 x 10.5 cm (101/4 x 65/16 x 4")
- Supplier No.:6902-2000
Spécifications
A propos de cet article
Pans are lightweight, durable, chemically inert, and autoclavable
For repeated autoclaving of small instruments and labware. Can also be used for soaking, cleaning, storing, or carrying general lab products. Heavy-duty construction with extra strong rims. Tapered for nesting.
Customers Who Bought This Also Bought