Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
# de catalogue 62662-026
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cat. no.:62662-026
- L×W×H:36 x 31 x 14 cm (143/16 x 127/32 x 51/2")
- Supplier No.:6900-0010
Spécifications
A propos de cet article
Useful for drying and sterilizing instruments and labware, and for collecting and washing soiled lab equipment
Rolled edges for extra stiffness. Handles for easy carrying. Tapered for nesting. Autoclavable.
Customers Who Bought This Also Bought