Order Entry
ContactUsLinkComponent
AKROBIN 14.75X16.5X7 BLUE
AkroBins® Bin Dividers and Hanging Systems, Akro-Mils™
Catalog # 62661-522
Supplier:  Akro-Mils
AKROBIN 14.75X16.5X7 BLUE
AkroBins® Bin Dividers and Hanging Systems, Akro-Mils™
Catalog # 62661-522
Supplier:  Akro-Mils

Specifications

  • Color:
    Blue
  • Description:
    Model 30250
  • Dimensions:
    37.5L x 42W x 17.8H cm (143/4 x 161/2 x 7")
  • Cat. no.:
    62661-522
  • Supplier No.:
    30250BLUE

Specifications

About this item

Heavy-duty polypropylene AkroBins® bins are waterproof, rustproof, corrosion-resistant, and impervious to weak acids and alkalis.

  • Autoclavable
  • Designed with an anti-slide stop, reinforced ribs, and a wide ridge for multi-level stacking

Clear polystyrene lids keep products clean, dust-free, and visible. Bin dividers maximize space for storage of smaller parts and accessories. Bin panel 62661-490 and bench rack 62661-498 are specifically designed to organize AkroBins® bins. The 16-gauge steel louvered bin panels with gray baked enamel finish are mountable on almost any vertical surface. The freestanding bench rack with gray baked enamel finish provides heavy-duty desktop storage. Bin panels and bench racks can be used with any size AkroBins® bins.

Ordering information: Bins, lids, dividers, and supports are ordered separately.
Customers Who Bought This Also Bought