Germicidal Ultraviolet Lamp, Labconco
# de catalogue 62403-141
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Power:30 W
- Weight:5 lbs.
- Voltage:115 V
- Supplier No.:1271300
Spécifications
A propos de cet article
This germicidal UV lamp is for use in Labconco PuriCare Procedure Stations (pre-2008), Purifier Cell Logic Biosafety Cabinets, Purifier Delta Class II Biosafety Cabinets, and Purifier Logic Class II Biosafety Cabinets.
Customers Who Bought This Also Bought