Specifications
- Format:Plate
- No. of wells:96
- Well Volume:0.28 mL
- Cat. no.:62402-954
- Well Shape:U-Bottom
- Supplier No.:3655
Specifications
About this item
These plates are ideal for a variety of colorimetric techniques, such as ELISA, which are performed in a 96-well format
Immulon 1 B exhibits a consistent medium binding affinity to proteins and complexes primarily with hydrophobic tendencies. Immulon 2 HB is an irradiated plate that provides higher binding affinity. Immulon 4 HBX features a unique resin that offers maximum protein uptake with enhanced precision and guaranteed coefficient of variation (CV) of <5%.
Choose from plates, Removawell™ Assemblies and Strips for custom formatting, and Dividastrips™ Assemblies with 2 x 8 strips that separate into two 1x8 strips.
Immulon polystyrene plastics provide a consistent surface for reproducible results over a variety of binding affinities.
Plate dimensions: 12.8 x 8.6 cm (5 x 33/8")